Learn More
SHANK3 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$151.31
Specifications
Antigen | SHANK3 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Liquid |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-156-7061
![]() |
Bethyl Laboratories, Inc
A304178AT |
20 μL |
Each of 1 for $151.31
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Rat
SH3 and multiple ankyrin repeat domains 3 (SHANK3), a member of the Shank gene family, plays a role in synapse formation and dendritic spine maturation. Shank proteins (Shank 1-3) containing PDZ domains are scaffold proteins of the postsynaptic density (PSD) that connect neurotransmitter receptors and ion channels proteins to the actin cytoskeleton and G-protein-coupled signaling pathways. Transcript splice variation in the Shank family influences the spectrum of Shank-interacting proteins in the PSDs of adult and developing brain to ensure normal development. Mutations of SHANK3 are a cause of autism spectrum disorder (ASD) and the neurological symptoms of 22q13.3 deletion syndrome.Specifications
SHANK3 | |
Polyclonal | |
Liquid | |
RUO | |
TBS with 0.1% BSA and 0.09% sodium azide | |
SHANK3, SH3 and multiple ankyrin repeat domains 3, ProSAP2, PSAP2, SPANK-2, Proline-rich synapse-associated protein 2 | |
Between 1475 and 1525 | |
Primary | |
4° C | |
SHANK3 |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
Q4ACU6, Q9BYB0 | |
58234, 85358 | |
IgG | |
Antigen affinity chromatography | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.