Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SHARPIN Antibody, Alexa Fluor™ 750, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP204116AF750
Description
SHARPIN Polyclonal antibody specifically detects SHARPIN in Human, Mouse samples. It is validated for Western BlotSpecifications
SHARPIN | |
Polyclonal | |
Western Blot | |
DKFZp434N1923, hSIPL1, SHANK-associated RH domain interactor, Shank-associated RH domain-interacting protein, Shank-interacting protein-like 1, sharpin, SIPL1DKFZP434N1923 | |
The immunogen recognized by this antibody maps to a region between residue 25 and 75 of human Shank-Associated RH Domain-Interacting Protein using the numbering given in entry NP_112236.3 (GeneID 81858). | |
0.1 mL | |
Primary | |
Human, Mouse | |
Purified |
Western Blot | |
Alexa Fluor 750 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
81858 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction