Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SHARPIN Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP204116
Description
SHARPIN Polyclonal specifically detects SHARPIN in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation.Specifications
SHARPIN | |
Polyclonal | |
Unconjugated | |
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
DKFZp434N1923, hSIPL1, SHANK-associated RH domain interactor, Shank-associated RH domain-interacting protein, Shank-interacting protein-like 1, sharpin, SIPL1DKFZP434N1923 | |
Rabbit | |
Affinity Purified | |
RUO | |
81858 | |
Human, Mouse | |
IgG |
Western Blot, Immunoprecipitation | |
1.0 mg/mL | |
Western Blot 1:2000-1:10000, Immunoprecipitation 2-10 mcg/mg | |
NP_112236.3 | |
SHARPIN | |
The immunogen recognized by this antibody maps to a region between residue 25 and 75 of human Shank-Associated RH Domain-Interacting Protein using the numbering given in entry NP_112236.3 (GeneID 81858). | |
100 μL | |
Primary | |
Based on 100% sequence identity, this antibody is predicted to react with Orangutan, Rhesus Monkey, Gorilla, Chimpanzee, Crab-eating macaque, European domestic ferret, Little brown bat and Northern white-cheeked gibbon. | |
Store at 4C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction