Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SHARPIN Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$181.90 - $402.00
Specifications
| Antigen | SHARPIN |
|---|---|
| Concentration | 0.5 mg/mL |
| Dilution | Western Blot 1:100-1:2000, Immunohistochemistry-Paraffin |
| Applications | Western Blot, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP227186SS
![]() |
Novus Biologicals
NBP227186SS |
0.025 mg |
Each for $181.90
|
|
|||||
NBP227186
![]() |
Novus Biologicals
NBP227186 |
0.1 mg |
Each for $402.00
|
|
|||||
Description
SHARPIN Polyclonal specifically detects SHARPIN in Human, Canine samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| SHARPIN | |
| Western Blot 1:100-1:2000, Immunohistochemistry-Paraffin | |
| Polyclonal | |
| Purified | |
| RUO | |
| 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide | |
| 81858 | |
| A portion of amino acids 25-75 of human Sharpin was used as the immunogen for this antibody. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 0.5 mg/mL | |
| Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Human, Canine | |
| DKFZp434N1923, hSIPL1, SHANK-associated RH domain interactor, Shank-associated RH domain-interacting protein, Shank-interacting protein-like 1, sharpin, SIPL1DKFZP434N1923 | |
| SHARPIN | |
| IgG | |
| Protein G purified | |
| The amino acid sequence used as immunogen for the antibody is 100% homologous in human and dog, 78% in mouse and 72% homologous in rat. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title