Learn More
Invitrogen™ SHARPIN Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA588439
Description
Positive Samples: 293T, LO2, HeLa, A-549, Rat brain; Cellular Location: Cell junction, Cytoplasm, cytosol, synapse Immunogen sequence: MAPPAGGAAA AASDLGSAAV LLAVHAAVRP LGAGPDAEAQ LRRLQLSADP ERPGRFRLEL LGAGPGAVNL EWPLESVSYT IRGPTQHELQ PPPGGPGTLS LHFLNPQEAQ RWAVLVRGAT VEGQNG.
Component of the LUBAC complex which conjugates linear polyubiquitin chains in a head-to-tail manner to substrates and plays a key role in NF-kappa-B activation and regulation of inflammation. LUBAC conjugates linear polyubiquitin to IKBKG and RIPK1 and is involved in activation of the canonical NF-kappa-B and the JNK signaling pathways. Linear ubiquitination mediated by the LUBAC complex interferes with TNF-induced cell death and thereby prevents inflammation. LUBAC is proposed to be recruited to the TNF-R1 signaling complex (TNF-RSC) following polyubiquitination of TNF-RSC components by BIRC2 and/or BIRC3 and to conjugate linear polyubiquitin to IKBKG and possibly other components contributing to the stability of the complex. Together with FAM105B/otulin, the LUBAC complex regulates the canonical Wnt signaling during angiogenesis.
Specifications
SHARPIN | |
Polyclonal | |
Unconjugated | |
SHARPIN | |
0610041B22Rik; AW121341; Conneck1; cpdm; hSIPL1; mSIPL1; protein kinase C-interacting protein RBCC like 1; PSEC0216; RBCKL1; SHANK associated RH domain interactor; SHANK-associated RH domain interacting protein; SHANK-associated RH domain interactor; shank-associated RH domain-interacting protein; shank-interacting protein; shank-interacting protein-like 1; SHARPIN; SIPL1 | |
Rabbit | |
Affinity chromatography | |
RUO | |
106025, 81858, 81859 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
2.08 mg/mL | |
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
Q91WA6, Q9EQL9, Q9H0F6 | |
SHARPIN | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-126 of human SHARPIN (NP_1122363?). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.