Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SHARPIN Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA540129
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA540129 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA540129 Supplier Invitrogen™ Supplier No. PA540129
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: K562 and THP-1 cells. Predicted reactivity: Chimpanzee (96%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Component of the LUBAC complex which conjugates linear polyubiquitin chains in a head-to-tail manner to substrates and plays a key role in NF-kappa-B activation and regulation of inflammation. LUBAC conjugates linear polyubiquitin to IKBKG and RIPK1 and is involved in activation of the canonical NF-kappa-B and the JNK signaling pathways. Linear ubiquitination mediated by the LUBAC complex interferes with TNF-induced cell death and thereby prevents inflammation. LUBAC is proposed to be recruited to the TNF-R1 signaling complex (TNF-RSC) following polyubiquitination of TNF-RSC components by BIRC2 and/or BIRC3 and to conjugate linear polyubiquitin to IKBKG and possibly other components contributing to the stability of the complex. Together with FAM105B/otulin, the LUBAC complex regulates the canonical Wnt signaling during angiogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SHARPIN
Applications Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 3.41 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Gene SHARPIN
Gene Accession No. Q9H0F6
Gene Alias 0610041B22Rik; AW121341; Conneck1; cpdm; hSIPL1; mSIPL1; protein kinase C-interacting protein RBCC like 1; PSEC0216; RBCKL1; SHANK associated RH domain interactor; SHANK-associated RH domain interacting protein; SHANK-associated RH domain interactor; shank-associated RH domain-interacting protein; shank-interacting protein; shank-interacting protein-like 1; SHARPIN; SIPL1
Gene Symbols SHARPIN
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human SHARPIN. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 81858
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.