Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SHB Antibody (SR1974), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP322624100UL
Description
SHB Monoclonal antibody specifically detects SHB in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
SHB | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot | |
SR1974 | |
Western Blot 1:500-1:2000 | |
bA3J10.2, RP11-3J10.8, SH2 domain-containing adapter protein B, SHB (Src homology 2 domain containing) adaptor protein B, SHB adaptor protein (a Src homology 2 protein), Src homology 2 domain containing adaptor protein B | |
A synthesized peptide derived from human SHB (Uniprot #: Q15464) | |
100 μg | |
Phospho Specific | |
6461 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction