Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SHBG Antibody (OTI1H10), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274166B
Description
SHBG Monoclonal specifically detects SHBG in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
SHBG | |
Monoclonal | |
Biotin | |
PBS with 0.05% Sodium Azide | |
SHBG | |
Full length recombinant protein of human SHBG (NP_001139751) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human | |
IgG2a |
Western Blot, Immunohistochemistry | |
OTI1H10 | |
Western Blot, Immunohistochemistry | |
ABPTestosterone-estrogen-binding globulin, androgen binding protein, MGC126834, MGC138391, SBP, sex hormone-binding globulin, Sex steroid-binding protein, TeBG, Testis-specific androgen-binding protein, testosterone-binding beta-globulin, Testosterone-estradiol-binding globulin | |
Mouse | |
Affinity Purified | |
Cancer, Vision | |
6462.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction