Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SHBG Antibody (SHBG/245), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP2449930.2MG
Description
Ensure accurate, reproducible results in Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence
SHBG Monoclonal antibody specifically detects SHBG in Human samples. It is validated for Flow Cytometry, Immunohistochemistry-Paraffin, SDS-Page, Immunofluorescence.Specifications
SHBG | |
Monoclonal | |
0.2mg/mL | |
Flow Cytometry 0.5 - 1 ug/million cells in 0.1 ml, Immunohistochemistry-Paraffin 1 - 2 ug/ml, SDS-Page, Immunofluorescence 0.5 - 1.0 ug/ml | |
P04278 | |
SHBG | |
Recombinant full-length human SHBG protein | |
Protein A or G purified | |
RUO | |
Primary | |
Recognizes a protein of 45kDa, identified as SHBG. It functions as an androgen transport protein, but may also be involved in receptor-mediated processes. Each dimer binds one molecule of steroid. It is specific for 5-alpha-dihydrotestosterone, testosterone, and 17-beta-estradiol. SHBG regulates the plasma metabolic clearance rate of steroid hormones by controlling their plasma concentration. In testis, it is synthesized by the Sertoli cells, secreted into the lumen of the seminiferous tubule and transported to the epididymis. | |
Store at 4C. | |
IgG1 |
Flow Cytometry, Immunohistochemistry (Paraffin), SDS-Page, Immunofluorescence | |
SHBG/245 | |
Unconjugated | |
10mM PBS and 0.05% BSA with 0.05% Sodium Azide | |
ABPTestosterone-estrogen-binding globulin, androgen binding protein, MGC126834, MGC138391, SBP, sex hormone-binding globulin, Sex steroid-binding protein, TeBG, Testis-specific androgen-binding protein, testosterone-binding beta-globulin, Testosterone-estradiol-binding globulin | |
Mouse | |
45 kDa | |
0.2 mg | |
Cancer, Vision | |
6462 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction