Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SHBG Monoclonal Antibody (H3), Invitrogen™
GREENER_CHOICE

Catalog No. FEMA551095
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA551095 200 μL
FEMA551096 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. FEMA551095 Supplier Invitrogen™ Supplier No. MA551095
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SHBG is a steroid binding protein that was first described as a plasma protein secreted by the liver and is thought to participate in the regulation of steroid responses. SHBG transports androgens and estrogens in the blood, binding each steroid molecule as a dimer formed from identical or nearly identical monomers. Low plasma SHBG levels are associated with obesity, abdominal adiposity, metabolic syndrome, and predict the development of type 2 diabetes. Polymorphisms in this gene have been associated with polycystic ovary syndrome and type 2 diabetes mellitus.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SHBG
Applications Western Blot, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone H3
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4
Gene SHBG
Gene Accession No. P04278
Gene Alias ABP; Abpa; androgen binding protein; Androgen binding protein (ABP); SBP; sex hormone binding globulin; Sex hormone binding globulin or androgen-binding protein (other gene product from ABP gene); sex hormone-binding globulin; Sex hormone-binding globulin (SHBG); Sex steroid binding protein (SBP); Sex steroid-binding protein; Shbg; TeBG; testis-specific androgen-binding protein; Testosterone binding beta globulin; testosterone-binding beta-globulin; testosterone-estradiol-binding globulin; Testosterone-Estradiol-Binding Globulin (TEBG); Testosterone-estrogen-binding globulin
Gene Symbols SHBG
Host Species Mouse
Immunogen Recombinant Human SHBG protein, Gly224-His388 (Accession #P04278).
Purification Method Protein A/G
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6462
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.