Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SHBG Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5112848
Description
SHBG Polyclonal Antibody for Western Blot, ELISA
SHBG is a steroid binding protein that was first described as a plasma protein secreted by the liver and is thought to participate in the regulation of steroid responses. SHBG transports androgens and estrogens in the blood, binding each steroid molecule as a dimer formed from identical or nearly identical monomers. Low plasma SHBG levels are associated with obesity, abdominal adiposity, metabolic syndrome, and predict the development of type 2 diabetes. Polymorphisms in this gene have been associated with polycystic ovary syndrome and type 2 diabetes mellitus.
Specifications
| SHBG | |
| Polyclonal | |
| Unconjugated | |
| SHBG | |
| ABP; Abpa; androgen binding protein; Androgen binding protein (ABP); SBP; sex hormone binding globulin; Sex hormone binding globulin or androgen-binding protein (other gene product from ABP gene); sex hormone-binding globulin; Sex hormone-binding globulin (SHBG); Sex steroid binding protein (SBP); Sex steroid-binding protein; Shbg; TeBG; testis-specific androgen-binding protein; Testosterone binding beta globulin; testosterone-binding beta-globulin; testosterone-estradiol-binding globulin; Testosterone-Estradiol-Binding Globulin (TEBG); Testosterone-estrogen-binding globulin | |
| Rabbit | |
| Protein G | |
| RUO | |
| 20415, 6462 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Western Blot | |
| 4.4 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| P04278, P97497 | |
| SHBG | |
| Recombinant Human Sex hormone-binding globulin protein (30-402AA). | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction