Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SHC Polyclonal Antibody
SHC Polyclonal Antibody
Supplier: Thermo Scientific PA579995
Description
SHC Polyclonal Antibody for Western Blot, ICC/IF, IHC (F), IHC (P)

Specifications
SHC | |
Polyclonal | |
Unconjugated | |
SHC1 | |
FLJ26504; OTTHUMP00000035472; p66; P66shc; RP11-307C12.1; SH2 domain protein C1; SHC; SHC (Src homology 2 domain containing) transforming protein 1; SHC (Src homology 2 domain-containing) transforming protein 1; SHC adaptor protein 1; SHC1; SHCA; SHC-transforming protein 1; SHC-transforming protein 3; SHC-transforming protein A; src homology 2 domain-containing transforming protein C1; src homology 2 domain-containing-transforming protein C1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
20416, 6464, 85385 | |
-20°C | |
Lyophilized |
Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
500 μg/mL | |
PBS with 5mg BSA and 0.05mg sodium azide, 0.05mg thimerosal | |
P29353, P98083, Q5M824 | |
SHC1 | |
A synthetic peptide corresponding to a sequence at the C-terminus of human SHC (424-440aa DPSYVNVQNLDKARQAV). | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction