Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SHC1 Antibody, HRP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP118908H
Description
SHC1 Polyclonal antibody specifically detects SHC1 in Human samples. It is validated for Western BlotSpecifications
SHC1 | |
Polyclonal | |
Western Blot | |
FLJ26504, p66, p66SHC, SH2 domain protein C1, SHC, SHC (Src homology 2 domain containing) transforming protein 1, SHC (Src homology 2 domain-containing) transforming protein 1, SHCA, SHC-transforming protein 1, SHC-transforming protein 3, SHC-transforming protein A, Src homology 2 domain-containing-transforming protein C1 | |
The immunogen recognized by this antibody maps to a region between residue 300 and 350 of human SHC (Src homology 2 domain containing) transforming protein 1 using the numbering given in entry NP_892113.4 (GeneID 6464). | |
0.1 mL | |
Cell Cycle and Replication, Phospho Specific, Signal Transduction | |
6464 | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
HRP | |
PBS | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction