Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SHC1 Antibody (OTI3A1) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274167
Description
SHC1 Monoclonal specifically detects SHC1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.Specifications
SHC1 | |
Monoclonal | |
LYOPH | |
Western Blot 1:2000, Immunohistochemistry 1:50, Immunocytochemistry/Immunofluorescence 1:100, Immunoprecipitation | |
FLJ26504, p66, p66SHC, SH2 domain protein C1, SHC, SHC (Src homology 2 domain containing) transforming protein 1, SHC (Src homology 2 domain-containing) transforming protein 1, SHCA, SHC-transforming protein 1, SHC-transforming protein 3, SHC-transforming protein A, Src homology 2 domain-containing-transforming protein C1 | |
Mouse | |
51.5 kDa | |
100 μg | |
Primary | |
Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended. | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG1 |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
OTI3A1 | |
Unconjugated | |
Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative | |
SHC1 | |
Full length human recombinant protein of human SHC1 (NP_003020) produced in HEK293T cell. | |
Affinity Purified | |
Cell Cycle and Replication, Phospho Specific, Signal Transduction | |
6464 | |
Human, Mouse, Rat | |
Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction