Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SHC2/Sck Rabbit anti-Human∣Mouse∣Rat, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS12144R

Description
Src homology 2 (SH2) domains bind specifically to tyrosine-phosphorylated proteins that temporally participate in signal transduction events. Shc-like protein (Sck) is a neuronal adaptor protein that contains an N-terminal PTB (phosphotyrosine binding) domain, a collagen homology (CH) domain, and a conserved C-terminal SH2 domain. Human Sck transcripts are present at high levels in liver, pancreas, prostate and ovary. In vascular endothelial cells, Sck participates in VEGF-induced signal transduction. Treatment of human umbilical vein endothelial (HUVEC) cells with VEGF induces recruitment of Sck to tyrosine-1175 of the kinase insert domain-containing receptor (KDR) and enhances Sck tyrosine phosphorylation.Specifications
SHC2/Sck | |
Polyclonal | |
Unconjugated | |
SHC2 | |
6720466E06; neuronal Shc adaptor homolog; Protein Sck; Protein Sli; Sck; SH2 domain protein C2; SHC (Src homology 2 domain containing) transforming protein 2; SHC adaptor protein 2; SHC2; ShcB; SHC-transforming protein 2; SHC-transforming protein B; Sli; src homology 2 domain-containing transforming protein C2; Src homology 2 domain-containing-transforming protein C2 | |
Rabbit | |
Protein A | |
RUO | |
216148, 25759, 314612 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
O70142, P98077, Q8BMC3 | |
SHC2 | |
KLH conjugated synthetic peptide derived from human SHC2/Sck. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction