Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SHC3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | SHC3 |
---|---|
Applications | Western Blot, Immunocytochemistry, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
SHC3 Polyclonal specifically detects SHC3 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
SHC3 | |
Polyclonal | |
Rabbit | |
Human | |
53358 | |
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:HPYYNSIPSKMPPPGGFLDTRLKPRPHAPDTAQFAGKEQTYYQGRHLGDTFGEDWQQTPLRQGSSDIYSTPEGKLHVAPTGEAPTYVNTQQIPP | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Western Blot, Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
RUO | |
Neuronal Shc, NSHCDKFZp686H1544, N-Shcsrc homology 2 domain containing transforming protein C3, Protein Rai, RAI, SH2 domain protein C3, SHC (Src homology 2 domain containing) transforming protein 3, SHCCFLJ45325, SHC-transforming protein 3, SHC-transforming protein C, src homology 2 domain-containing transforming protein C3, Src homology 2 domain-containing-transforming protein C3 | |
SHC3 | |
IgG | |
Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title