Learn More
Invitrogen™ SHC4 Polyclonal Antibody

Description
SHC4 Polyclonal Antibody for Western Blot
Specifications
Specifications
| Antigen | SHC4 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.25 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | SHC4 |
| Gene Accession No. | Q6S5L9 |
| Gene Alias | 6230417E10Rik; 9930029B02Rik; Gm685; hShcD; mShcD; novel phosphotyrosine interaction domain (PTB/PID) and SH2 domains containing protein; rai-like protein; rai-like protein RaLP; RaLP; RGD1309182; SH2 domain protein C4; SHC (Src homology 2 domain containing) family member 4; SHC (Src homology 2 domain containing) family, member 4; SHC adaptor protein 4; Shc4; Shcd; SHC-transforming protein 4; SHC-transforming protein D; src homology 2 domain-containing-transforming protein C4; UNQ6438/PRO21364 |
| Gene Symbols | SHC4 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.