Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SHC4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5113027
Description
SHC4 Polyclonal Antibody for Western Blot, IHC (P), ELISA
Activates both Ras-dependent and Ras-independent migratory pathways in melanomas. Contributes to the early phases of agrin-induced tyrosine phosphorylation of CHRNB1.
Specifications
| SHC4 | |
| Polyclonal | |
| Unconjugated | |
| SHC4 | |
| 6230417E10Rik; 9930029B02Rik; Gm685; hShcD; mShcD; novel phosphotyrosine interaction domain (PTB/PID) and SH2 domains containing protein; rai-like protein; rai-like protein RaLP; RaLP; RGD1309182; SH2 domain protein C4; SHC (Src homology 2 domain containing) family member 4; SHC (Src homology 2 domain containing) family, member 4; SHC adaptor protein 4; Shc4; Shcd; SHC-transforming protein 4; SHC-transforming protein D; src homology 2 domain-containing-transforming protein C4; UNQ6438/PRO21364 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 399694 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 0.73 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q6S5L8 | |
| SHC4 | |
| Recombinant Human SHC-transforming protein 4 protein (1-185AA). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction