Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SHC4 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501979980
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-197-9980 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-197-9980 Supplier Bioss Supplier No. BS8700R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Activates both Ras-dependent and Ras-independent migratory pathways in melanomas. Contributes to the early phases of agrin-induced tyrosine phosphorylation of CHRNB1.

Specifications

Antigen SHC4
Applications Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA and 15mM sodium azide; pH
Gene SHC4
Gene Accession No. Q6S5L8
Gene Alias 6230417E10Rik; 9930029B02Rik; Gm685; hShcD; mShcD; novel phosphotyrosine interaction domain (PTB/PID) and SH2 domains containing protein; Rai-like protein; rai-like protein RaLP; RaLP; SH2 domain protein C4; SHC (Src homology 2 domain containing) family member 4; SHC (Src homology 2 domain containing) family, member 4; SHC adaptor protein 4; Shc4; SHCD; SHC-transforming protein 4; SHC-transforming protein D; Src homology 2 domain-containing-transforming protein C4; UNQ6438/PRO21364
Gene Symbols SHC4
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human SHC4.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 399694
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.