Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SHCBP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561827
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561827 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561827 Supplier Invitrogen™ Supplier No. PA561827
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VEGLKLYSEM EQLKQKLKLI ENPLLRYVFG YQKNSNIQAK GVRSSGQKIT HVVSSTMMAG LLRSLLTDRL CQEPGEEERE IQFHSDPLSA IN Highest antigen sequence identity to the following orthologs: Mouse - 77%, Rat - 79%.

May play a role in signaling pathways governing cellular proliferation, cell growth and differentiation. May be a component of a novel signaling pathway downstream of Shc. Acts as a positive regulator of FGF signaling in neural progenitor cells. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen SHCBP1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Shcbp1
Gene Accession No. Q8NEM2
Gene Alias mPAL; Pal; Protein expressed in Activated Lymphocytes; SHC binding and spindle associated 1; SHC SH2 domain-binding protein 1; SHC SH2-domain binding protein 1; SHC-binding protein; Shcbp1
Gene Symbols Shcbp1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SHCBP1. Recombinant protein control fragment (Product #RP-98589).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79801
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.