Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SHCBP1L Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA564183
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA564183 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA564183 Supplier Invitrogen™ Supplier No. PA564183
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TLRGVNMKVL PAPKLKMTNN HIYSNKGYGV SILQPMEQFF IVAEEALNKR ASSGDKKDDK MLFKVMQNLN LEMNNNKI Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 86%.

This gene encodes a Src homology 2 domain-binding protein 1-like protein. The encoded protein interacts with heat shock 70 kDa protein 2 and may be involved in maintaining spindle integrity during meiosis. This gene is located in region of chromoso0me 1 encompassing a prostate cancer susceptibility locus.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SHCBP1L
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SHCBP1L
Gene Accession No. Q9BZQ2
Gene Alias 1700012A16Rik; C1orf14; GE36; RGD1305609; SHC binding and spindle associated 1 like; SHC SH2 domain-binding protein 1-like protein; SHC SH2-domain binding protein 1-like; SHCBP1L; Testicular spindle-associated protein SHCBP1L
Gene Symbols SHCBP1L
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SHCBP1L. Recombinant protein control fragment (Product #RP-103571).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 81626
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.