Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SHCBP1L Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA569995
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA569995 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA569995 Supplier Invitrogen™ Supplier No. PA569995
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Human: 100%.

This gene encodes a Src homology 2 domain-binding protein 1-like protein. The encoded protein interacts with heat shock 70 kDa protein 2 and may be involved in maintaining spindle integrity during meiosis. This gene is located in region of chromoso0me 1 encompassing a prostate cancer susceptibility locus.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SHCBP1L
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene SHCBP1L
Gene Accession No. Q9BZQ2
Gene Alias 1700012A16Rik; C1orf14; GE36; RGD1305609; SHC binding and spindle associated 1 like; SHC SH2 domain-binding protein 1-like protein; SHC SH2-domain binding protein 1-like; SHCBP1L; Testicular spindle-associated protein SHCBP1L
Gene Symbols SHCBP1L
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human SHCBP1L (aa 13-62).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 81626
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.