Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SHD Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$243.50 - $670.50

Specifications

Antigen SHD
Dilution Western Blot 1:500, ELISA 1:10000
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB018751
SDP
View Documents
Novus Biologicals
NBP1781360.025MG
0.025 mg
Each for $243.50
Only null left
Add to Cart
 
NBP17813605
SDP
View Documents
Novus Biologicals
NBP1781360.05MG
0.05 mg
Each for $402.00
Only null left
Add to Cart
 
NBP178136
SDP
View Documents
Novus Biologicals
NBP178136
0.1 mg
Each for $670.50
Only null left
Add to Cart
 
Description

Description

SHD Polyclonal specifically detects SHD in Drosophila samples. It is validated for Western Blot, ELISA.
Specifications

Specifications

SHD
ELISA, Western Blot
Unconjugated
Drosophila
SH2 domain-containing adapter protein D, Src homology 2 domain containing transforming protein D, src homology 2 domain-containing transforming protein D
SHD
IgG
Western Blot 1:500, ELISA 1:10000
Polyclonal
Rabbit
Q9VUF8
56961
Synthetic peptide taken form the Spook protein form within the N-terminal region (amino acid 51-60) the peptide was post-synthetically modified and conjugated to carrier protein to achieve desired antigenicity. The Shade peptides sequences are conserved various other specieis of fruit flys.
Affinity Purified
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.