Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SHE Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557944
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557944 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557944 Supplier Invitrogen™ Supplier No. PA557944
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PYDAQQMITE IRRRGSKDPL VKALQLLDSP CEPADGGLKS ETLAKRRSSK DLLGKPPQLY DTPY Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 86%.

SHE, also known as SH2 domain-containing adapter protein E, is a 495 amino acid protein whose molecular weight is 54 kDa. The function of SHE remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SHE
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SHE
Gene Accession No. Q5VZ18
Gene Alias 9430022A14; RGD1563897; SH2 domain-containing adapter protein E; SHE; Src homology 2 domain containing E; src homology 2 domain-containing transforming protein E
Gene Symbols SHE
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SHE. Recombinant protein control fragment (Product #RP-96523).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 126669
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.