Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SHE Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113000
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113000 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113000 Supplier Invitrogen™ Supplier No. PA5113000
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SHE Polyclonal Antibody for Western Blot, IHC (P), ELISA

SHE, also known as SH2 domain-containing adapter protein E, is a 495 amino acid protein whose molecular weight is 54 kDa. The function of SHE remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SHE
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 3.31 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene SHE
Gene Accession No. Q5VZ18, Q8BSD5
Gene Alias 9430022A14; RGD1563897; SH2 domain-containing adapter protein E; SHE; Src homology 2 domain containing E; src homology 2 domain-containing transforming protein E
Gene Symbols SHE
Host Species Rabbit
Immunogen Recombinant Human SH2 domain-containing adapter protein E protein (2-162AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 126669, 214547, 685088
Target Species Human, Mouse, Rat
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.