Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SHE Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5114331
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5114331 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5114331 Supplier Invitrogen™ Supplier No. PA5114331
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SHE Polyclonal Antibody for IHC (P), Western Blot

SHE, also known as SH2 domain-containing adapter protein E, is a 495 amino acid protein whose molecular weight is 54 kDa. The function of SHE remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SHE
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4
Gene SHE
Gene Accession No. 0
Gene Alias 9430022A14; RGD1563897; SH2 domain-containing adapter protein E; SHE; Src homology 2 domain containing E; src homology 2 domain-containing transforming protein E
Gene Symbols SHE
Host Species Rabbit
Immunogen Recombinant protein within zebrafish SHE aa 64-291.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 561493
Target Species Zebrafish
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.