Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
17 alpha-Boldenone, Sheep, Polyclonal Antibody, Abnova™
Sheep polyclonal antibody raised against 17 alpha-Boldenone.
Supplier: Abnova Corporation PAB8199
Specifications
17 alpha-Boldenone | |
Polyclonal | |
Sheep polyclonal antibody raised against 17 alpha-Boldenone. | |
In 20mM phosphate buffer, 150mM NaCl, pH 7.2 (0.09% sodium azide) | |
17 alpha-Boldenone conjugated with BTG. | |
RUO | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
ELISA | |
Unconjugated | |
Enzyme Immunoassay (0.25 ug/mL) The optimal working dilution should be determined by the end user. | |
Sheep | |
100 μL | |
Primary | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction