Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Adarb1, Sheep, Polyclonal Antibody, Abnova™

Catalog No. 89114804 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-114-804 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-114-804 Supplier Abnova Corporation Supplier No. PAB10359
Add to Cart
Add to Cart

Sheep polyclonal antibody raised against partial recombinant Adarb1.

Specifications

Antigen Adarb1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Sheep polyclonal antibody raised against partial recombinant Adarb1.
Dilution Western blot (1:1000) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.08% sodium azide)
Gene Adarb1
Gene Alias Adar2/Red1
Gene Symbols Adarb1
Host Species Sheep
Immunogen Recombinant GST fusion protein corresponding to amino acids 620-1597 of rat Adarb1.
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25367
Target Species Rat
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.