Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AUP1, Sheep, Polyclonal Antibody, Abnova™

Catalog No. 89114398 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-114-398 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-114-398 Supplier Abnova Corporation Supplier No. PAB0753
Add to Cart
Add to Cart

Sheep polyclonal antibody raised against synthetic peptide of AUP1.

This gene encodes a protein that contains a domain with homology to the ancient conserved region of the archain 1 gene and a domain that may be involved in binding ubiquitin-conjugating enzymes. The protein encoded by this gene has been shown to bind to the conserved membrane-proximal sequence of the cytoplasmic tail of integrin alpha(IIb) subunits. These subunits play a crucial role in the integrin alpha(IIb)beta(3) inside-out signalling in platelets and megakaryocytes that leads to platelet aggregation and thrombus formation. This gene overlaps the gene for mitochondrial serine protease 25. [provided by RefSeq

Sequence: HVFLVSCALPDSV

Specifications

Antigen AUP1
Applications Immunohistochemistry (Frozen), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Sheep polyclonal antibody raised against synthetic peptide of AUP1.
Dilution Immunohistochemistry (1:500-1:6000) Western Blot (1:500-1:6000) The optimal dilution should be determined by the end user.
Formulation Lyophilized from PBS
Gene AUP1
Gene Accession No. Q9Y679;Q3ZC65;P70295;A1L134
Gene Symbols AUP1
Host Species Sheep
Immunogen A synthetic peptide (conjugated with Blue Carrier Protein) corresponding to amino acids 48-60 of human AUP1.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 550
Test Specificity This antiserum is specific to ancient ubiquitous protein 1.
Target Species Bovine, Human, Rat, Sheep
Content And Storage Store at 4°C on dry atmosphere. After reconstitution with deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.