Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AURKB, Sheep, Polyclonal Antibody, Abnova™

Catalog No. 89115743 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-115-743 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-115-743 Supplier Abnova Corporation Supplier No. PAB12360
Add to Cart
Add to Cart

Sheep polyclonal antibody raised against partial recombinant AURKB.

Chromosomal segregation during mitosis as well as meiosis is regulated by kinases and phosphatases. The Aurora kinases associate with microtubules during chromosome movement and segregation. Aurora kinase B localizes to microtubules near kinetochores, specifically to the specialized microtubules called K-fibers, and Aurora kinase A (MIM 603072) localizes to centrosomes (Lampson et al., 2004 [PubMed 14767480]).[supplied by OMIM

Specifications

Antigen AURKB
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Sheep polyclonal antibody raised against partial recombinant AURKB.
Dilution Western Blot (0.2 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.09% sodium azide)
Gene AURKB
Gene Alias AIK2/AIM-1/AIM1/ARK2/AurB/IPL1/STK12/STK5
Gene Symbols AURKB
Host Species Sheep
Immunogen Recombinant His6 fusion protein corresponding to amino acids 1-344 of human AURKB.
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9212
Test Specificity recognizes human AURKB.
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.