Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bovine Serum Albumin, BSA Sheep, Polyclonal Antibody, Abnova™

Catalog No. 89136914 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-136-914 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-136-914 Supplier Abnova Corporation Supplier No. PAB0903
Add to Cart
Add to Cart

Sheep polyclonal antibody raised against Bovine Serum Albumin, BSA.

Specifications

Antigen Bovine Serum Albumin, BSA
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Sheep polyclonal antibody raised against Bovine Serum Albumin, BSA.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS (0.2% gelatin, 0.09% sodium azide)
Host Species Sheep
Immunogen Bovine Serum Albumin, BSA.
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bovine
Content And Storage Store in the dark at 4°C. Do not freeze.
Avoid prolonged exposure to light.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.