Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bub1b Sheep anti-Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89134413 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
250 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-134-413 250 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-134-413 Supplier Abnova Corporation Supplier No. PAB9784
Add to Cart
Add to Cart

Sheep polyclonal antibody raised against partial recombinant Bub1b.

Specifications

Antigen Bub1b
Applications Immunohistochemistry, Immunoprecipitation
Classification Polyclonal
Conjugate Unconjugated
Description Sheep polyclonal antibody raised against partial recombinant Bub1b.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS (0.08% sodium azide)
Gene Bub1b
Gene Alias AU045529/BUBR1/MGC36092
Gene Symbols Bub1b
Host Species Sheep
Immunogen Recombinant protein corresponding to amino acids 10-190 of mouse Bub1b.
Quantity 250 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12236
Target Species Mouse
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.