Learn More
CALCA, Sheep, Polyclonal Antibody, Abnova™
Sheep polyclonal antibody raised against partial length recombinant CALCA.
Supplier: Abnova Corporation PAB8431
Description
Calcitonin is a peptide hormone synthesized by the parafollicular cells of the thyroid. It causes reduction in serum calcium, an effect opposite to that of parathyroid hormone (PTH; MIM 168450).[supplied by OMIM
Specifications
CALCA | |
Polyclonal | |
Sheep polyclonal antibody raised against partial length recombinant CALCA. | |
In 20mM phosphate buffer, 150mM NaCl, pH 7.2 (0.09% sodium azide) | |
CALC1/CGRP/CGRP-I/CGRP1/CT/KC/MGC126648 | |
Sheep | |
1 mL | |
Primary | |
Human | |
Liquid |
ELISA | |
Unconjugated | |
The optimal working dilution should be determined by the end user. | |
CALCA | |
CALCA | |
Recombinant His fusion protein corresponding to amino acids 3-116 of human CALCA. | |
RUO | |
796 | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only