Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytochrome c, Sheep, Polyclonal Antibody, Abnova™

Catalog No. 89139144 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-139-144 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-139-144 Supplier Abnova Corporation Supplier No. PAB9852
Add to Cart
Add to Cart

Sheep polyclonal antibody raised against full-length recombinant Cytochrome c.

Specifications

Antigen Cytochrome c
Applications Immunofluorescence, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Sheep polyclonal antibody raised against full length recombinant Cytochrome c.
Dilution Western Blot (1:5000) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.08% sodium azide)
Host Species Sheep
Immunogen Recombinant protein corresponding to full length Cytochrome c.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Canine, Human, Rabbit, Rat
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.