Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

dynamin (phospho S774), Sheep, Polyclonal Antibody, Abnova™

Catalog No. 89139137 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-139-137 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-139-137 Supplier Abnova Corporation Supplier No. PAB9697
Add to Cart
Add to Cart

Sheep polyclonal antibody raised against synthetic phosphopeptide of dynamin.

Specifications

Antigen dynamin
Applications Immunofluorescence, Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Sheep polyclonal antibody raised against synthetic phosphopeptide of dynamin.
Dilution Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:1000) Immunofluorescence (1:1000) Western Blot (1:1000) The optimal working dilution should be determined by the end user.
Formulation In 10mM HEPES, 150mM NaCl, pH 7.5 (50% glycerol, 10% BSA)
Gene Accession No. P21575
Host Species Sheep
Immunogen Synthetic phosphopeptide corresponding to residues surrounding S774 of rat dynamin.
Purification Method Affinity purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bovine, Canine, Chicken, Human, Mouse, Primate, Rat
Content And Storage Store at -80°C. For short term storage, store at -20°C. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.