Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

F10 Sheep anti-Mouse, Rat, Polyclonal Antibody, Abnova™

Catalog No. 89118844 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-118-844 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-118-844 Supplier Abnova Corporation Supplier No. PAB17094
Add to Cart
Add to Cart

Sheep polyclonal antibody raised against F10.

Specifications

Antigen F10
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Sheep polyclonal antibody raised against F10.
Dilution ELISA (10 ug/mL) Western Blot (10 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (50% glycerol)
Gene F10
Gene Alias Cf10/fX
Gene Symbols F10
Host Species Sheep
Immunogen Purified mouse F10.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14058
Target Species Mouse, Rat
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.