Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

F10 Sheep anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89117350 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-117-350 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-117-350 Supplier Abnova Corporation Supplier No. PAB14923
Add to Cart
Add to Cart

Sheep polyclonal antibody raised against F10.

This gene encodes the vitamin K-dependent coagulation factor X of the blood coagulation cascade. This factor undergoes multiple processing steps before its preproprotein is converted to a mature two-chain form by the excision of the tripeptide RKR. Two chains of the factor are held together by 1 or more disulfide bonds; the light chain contains 2 EGF-like domains, while the heavy chain contains the catalytic domain which is structurally homologous to those of the other hemostatic serine proteases. The mature factor is activated by the cleavage of the activation peptide by factor IXa (in the intrisic pathway), or by factor VIIa (in the extrinsic pathway). The activated factor then converts prothrombin to thrombin in the presence of factor Va, Ca+2, and phospholipid during blood clotting. Mutations of this gene result in factor X deficiency, a hemorrhagic condition of variable severity. [provided by RefSeq]

Specifications

Antigen F10
Applications ELISA, Immunocytochemistry, Immunodiffusion, Immunofluorescence, Immunomicroscopy
Classification Polyclonal
Conjugate Unconjugated
Description Sheep polyclonal antibody raised against F10.
Dilution ELISA (1:1000) Immunodiffusion (1:32) The optimal working dilution should be determined by the end user.
Formulation In serum
Gene F10
Gene Alias FX/FXA
Gene Symbols F10
Host Species Sheep
Immunogen Purified human F10.
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2159
Test Specificity This antibody has a single antigen reactivity with minimal cross reactivity with other human serum proteins.
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.