Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

F2 Sheep anti-Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89114337 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-114-337 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-114-337 Supplier Abnova Corporation Supplier No. PAB0685
Add to Cart
Add to Cart

Sheep polyclonal antibody raised against native F2.

Specifications

Antigen F2
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Sheep polyclonal antibody raised against native F2.
Dilution Western Blot (25 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (50% glycerol)
Gene F2
Gene Alias Cf-2/Cf2/FII
Gene Symbols F2
Host Species Sheep
Immunogen Native purified Mouse F2.
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14061
Test Specificity This antibody recognizes mouse prothrombin.
Target Species Mouse
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.