Learn More
F5 Sheep anti-Human, Polyclonal Antibody, Abnova™
Sheep polyclonal antibody raised against native F5.
Supplier: Abnova Corporation PAB0664
Description
This gene encodes an essential cofactor of the blood coagulation cascade. This factor circulates in plasma, and is converted to the active form by the release of the activation peptide by thrombin during coagulation. This generates a heavy chain and a light chain which are held together by calcium ions. The activated protein is a cofactor that participates with activated coagulation factor X to activate prothrombin to thrombin. Defects in this gene result in either an autosomal recessive hemorrhagic diathesis or an autosomal dominant form of thrombophilia, which is known as activated protein C resistance. [provided by RefSeq
Specifications
F5 | |
Polyclonal | |
Sheep polyclonal antibody raised against native F5. | |
In PBS (50% glycerol) | |
FVL/PCCF | |
Sheep | |
1 mg | |
Primary | |
This antibody recognizes human factor V. | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
ELISA, Western Blot | |
Unconjugated | |
Western Blot (25 ug/mL) The optimal working dilution should be determined by the end user. | |
F5 | |
F5 | |
Native purified Human F5. | |
RUO | |
2153 | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only