Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FGF2 Sheep anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™

Catalog No. 89114393 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-114-393 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-114-393 Supplier Abnova Corporation Supplier No. PAB0746
Add to Cart
Add to Cart

Sheep polyclonal antibody raised against full-length recombinant FGF2.

The protein encoded by this gene is a member of the fibroblast growth factor (FGF) family. FGF family members bind heparin and possess broad mitogenic and angiogenic activities. This protein has been implicated in diverse biological processes, such as limb and nervous system development, wound healing, and tumor growth. The mRNA for this gene contains multiple polyadenylation sites, and is alternatively translated from non-AUG (CUG) and AUG initiation codons, resulting in five different isoforms with distinct properties. The CUG-initiated isoforms are localized in the nucleus and are responsible for the intracrine effect, whereas, the AUG-initiated form is mostly cytosolic and is responsible for the paracrine and autocrine effects of this FGF. [provided by RefSeq

Specifications

Antigen FGF2
Applications Immunohistochemistry (Frozen), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Sheep polyclonal antibody raised against full length recombinant FGF2.
Dilution Immunohistochemistry (1:1000-1:2000) Western Blot (1:1000-1:2000) The optimal working dilution should be determined by the end user.
Formulation Lyophilized from PBS
Gene FGF2
Gene Accession No. P09038
Gene Alias BFGF/FGFB/HBGF-2
Gene Symbols FGF2
Host Species Sheep
Immunogen Recombinant protein corresponding to full length human FGF2.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2247
Test Specificity A high level of specificity for bFGF was shown by immunohistochemistry for this antiserum.
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C on dry atmosphere. After reconstitution with deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.