Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FOS Sheep anti-Hamster, Human, Rabbit, Rat, Polyclonal Antibody, Abnova™

Catalog No. 89114397 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-114-397 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-114-397 Supplier Abnova Corporation Supplier No. PAB0752
Add to Cart
Add to Cart

Sheep polyclonal antibody raised against synthetic peptide of FOS.

The Fos gene family consists of 4 members: FOS, FOSB, FOSL1, and FOSL2. These genes encode leucine zipper proteins that can dimerize with proteins of the JUN family, thereby forming the transcription factor complex AP-1. As such, the FOS proteins have been implicated as regulators of cell proliferation, differentiation, and transformation. In some cases, expression of the FOS gene has also been associated with apoptotic cell death. [provided by RefSeq

Sequence: MFSGFNADYEASSSRC

Specifications

Antigen FOS
Applications Immunohistochemistry (Frozen), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Sheep polyclonal antibody raised against synthetic peptide of FOS.
Dilution Immunohistochemistry (1:500-1:8000) Western Blot (1:500-1:8000) The optimal dilution should be determined by the end user.
Formulation Lyophilized from PBS
Gene FOS
Gene Accession No. P01100;Q4FDN1;P01101;Q56TL8;Q5XQC6
Gene Alias AP-1/C-FOS
Gene Symbols FOS
Host Species Sheep
Immunogen A synthetic peptide (conjugated diphtheria toxoid) corresponding to amino acids 2-17 of human FOS.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2353
Test Specificity This antiserum shows a high level of specificity for cFOS confirmed by immunohostochemistry.
Target Species Hamster, Human, Rabbit, Rat
Content And Storage Store at 4°C on dry atmosphere. After reconstitution with 100μL of deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.