Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gja5, Sheep, Polyclonal Antibody, Abnova™

Catalog No. 89114405 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-114-405 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-114-405 Supplier Abnova Corporation Supplier No. PAB0764
Add to Cart
Add to Cart

Sheep polyclonal antibody raised against synthetic peptide of Gja5.

Sequence: SLVQGLTPPPDFNQCLK

Specifications

Antigen Gja5
Applications Immunofluorescence, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Sheep polyclonal antibody raised against synthetic peptide of Gja5.
Dilution Immunofluorescence (1:10000-1:50000) Western Blot (1:10000-1:50000) The optimal working dilution should be determined by the end user.
Formulation Lyophilized from PBS
Gene Gja5
Gene Accession No. P28234
Gene Alias Cx40
Gene Symbols Gja5
Host Species Sheep
Immunogen A synthetic peptide (conjugated diphtheria toxoid) corresponding to amino acids 253-269 of rat Gja5.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 50563
Test Specificity Western blotting of cell membarnes of COS cells transiently transfected with cDNA encoding Connexin-40 shows two bands of molecular weight 38 and 40 KDa which are abolished by preincubation with the immunising peptide. No such bands are detected in western blots of membranes from non-transfected cells or cells transfected with cDNA encoding Connexin-37, Connexin-43 or Connexin-45.
Target Species Rat
Content And Storage Store at 4°C on dry atmosphere. After reconstitution with deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.