Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glucocorticoid receptor, Sheep, Polyclonal Antibody, Abnova™

Catalog No. 89138296 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-138-296 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-138-296 Supplier Abnova Corporation Supplier No. PAB17112
Add to Cart
Add to Cart

Sheep polyclonal antibody raised against synthetic peptide of Glucocorticoid receptor.

Specifications

Antigen Glucocorticoid receptor
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Sheep polyclonal antibody raised against synthetic peptide of Glucocorticoid receptor.
Dilution Western Blot (1:500-1:1000) The optimal working dilution should be determined by the end user.
Formulation In serum
Host Species Sheep
Immunogen A synthetic peptide (conjugated with diptheria toxoid) corresponding to amino acids 150-175 of squirrel monkey Glucocorticoid receptor.
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This antibody reacts specifically with monkey and human glucocorticoid receptor. This sequence is identical in human glucocorticoid receptor except for 2 amino acids (underlined): N terminus - APKEKEFPKTHSDVSSEQQNLKGQTG-NH2 - C terminus.
Target Species Human, Monkey, Squirrel
Content And Storage Store at -20°C. For long term storage store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.