Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Herpes Simplex Virus Type-2, Sheep, FITC, Polyclonal Antibody, Abnova™

Catalog No. 89137892 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-137-892 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-137-892 Supplier Abnova Corporation Supplier No. PAB14261
Add to Cart
Add to Cart

Sheep polyclonal antibody raised against Herpes Simplex Virus Type-2.

Specifications

Antigen Herpes Simplex Virus Type-2
Applications ELISA, Immunofluorescence, Western Blot
Classification Polyclonal
Conjugate FITC
Description Sheep polyclonal antibody raised against Herpes Simplex Virus Type-2.
Dilution The optimal working dilution should be determined by the end user.
Formulation In 10mM PBS, pH 7.2 (10mg/mL BSA, 0.09% sodium azide)
Host Species Sheep
Immunogen Native from human HSV-2, Strain G.
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity ICPs + late structural (virion) antigens. Does not react with HEp-2 cells. Has moderate cross reactivity with HSV type 1.
Target Species Herpes Simplex virus (HSV)
Content And Storage Store in the dark at 4°C. For long term storage store at -20°C.
Avoid prolonged exposure to light.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.