Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Herpes Simplex Virus Type-2, Sheep, HRP, Polyclonal Antibody, Abnova™

Catalog No. 89137893 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-137-893 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-137-893 Supplier Abnova Corporation Supplier No. PAB14262
Add to Cart
Add to Cart

Sheep polyclonal antibody raised against Herpes Simplex Virus Type-2.

Specifications

Antigen Herpes Simplex Virus Type-2
Applications ELISA, Immunocytochemistry, Western Blot
Classification Polyclonal
Conjugate HRP
Description Sheep polyclonal antibody raised against Herpes Simplex Virus Type-2.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS (10mg/mL BSA, 0.002% thimerosal)
Host Species Sheep
Immunogen Native from human HSV-2, Strain G.
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity ICPs + late structural (virion) antigens. Cross-reacts with HSV Type 1 titer is 1:512 by Indirect IF. Does not react with HEp-2 cells.
Target Species Herpes Simplex virus (HSV)
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.