Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

herpes virus 8 minor capsid protein, Sheep, Polyclonal Antibody, Abnova™

Catalog No. 89136973 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
250 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-136-973 250 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-136-973 Supplier Abnova Corporation Supplier No. PAB10423
Add to Cart
Add to Cart

Sheep polyclonal antibody raised against partial recombinant human herpes virus 8 minor capsid protein.

Specifications

Antigen herpes virus 8 minor capsid protein
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Sheep polyclonal antibody raised against partial recombinant human herpes virus 8 minor capsid protein.
Dilution Western blot (1:500) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.08% sodium azide)
Host Species Sheep
Immunogen Recombinant protein corresponding to N-terminus the maltose binding protein and C- terminal the protein encoded by the open reading frame 62 (ORF62) of human herpes virus 8 minor capsid protein.
Quantity 250 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.