Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL2, Sheep, Polyclonal Antibody, Abnova™

Catalog No. 89133641 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-133-641 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-133-641 Supplier Abnova Corporation Supplier No. PAB8417
Add to Cart
Add to Cart

Sheep polyclonal antibody raised against partial length recombinant IL2.

The protein encoded by this gene is a secreted cytokine that is important for the proliferation of T and B lymphocytes. The receptor of this cytokine is a heterotrimeric protein complex whose gamma chain is also shared by interleukin 4 (IL4) and interleukin 7 (IL7). The expression of this gene in mature thymocytes is monoallelic, which represents an unusual regulatory mode for controlling the precise expression of a single gene. The targeted disruption of a similar gene in mice leads to ulcerative colitis-like disease, which suggests an essential role of this gene in the immune response to antigenic stimuli. [provided by RefSeq

Specifications

Antigen IL2
Applications ELISA
Classification Polyclonal
Conjugate Unconjugated
Description Sheep polyclonal antibody raised against partial length recombinant IL2.
Dilution The optimal working dilution should be determined by the end user.
Formulation In 20mM phosphate buffer, 150mM NaCl, pH 7.2 (0.09% sodium azide)
Gene IL2
Gene Alias IL-2/TCGF/lymphokine
Gene Symbols IL2
Host Species Sheep
Immunogen Recombinant His fusion protein corresponding to amino acids 21-153 of human IL2.
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3558
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.