Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NEK2, Sheep, Polyclonal Antibody, Abnova™

Catalog No. 89115734 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-115-734 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-115-734 Supplier Abnova Corporation Supplier No. PAB12347
Add to Cart
Add to Cart

Sheep polyclonal antibody raised against synthetic peptide of NEK2.

Sequence: NLERRGRQLGEPEK

Specifications

Antigen NEK2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Sheep polyclonal antibody raised against synthetic peptide of NEK2.
Dilution Western Blot (0.5 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.09% sodium azide)
Gene NEK2
Gene Alias HsPK21/NEK2A/NLK1
Gene Symbols NEK2
Host Species Sheep
Immunogen A synthetic peptide corresponding to amino acids 282-295 of human NEK2.
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4751
Test Specificity recognizes both splice variants of human NEK2 (NEK2A and NEK2B).
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.