Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ngf Sheep anti-Birds, Human, Mouse, Rat, Polyclonal Antibody, Abnova™

Catalog No. 89114400 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
250 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-114-400 250 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-114-400 Supplier Abnova Corporation Supplier No. PAB0755
Add to Cart
Add to Cart

Sheep polyclonal antibody raised against native Ngf.

Specifications

Antigen Ngf
Applications Dot Blot, ELISA, Immunohistochemistry (Frozen), Inhibition Assays, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Sheep polyclonal antibody raised against native Ngf.
Dilution Immunohistochemistry (1:1000-1:5000) ELISA (1:15000) Western Blot (1:1000-1:5000) Dot Blot (1:1000-1:5000) Inhibition assay (1:10-50 for in vitro 5-10 ul/g body weight for in vivo) The optimal working dilution should be determined by the
Formulation Lyophilized from PBS
Gene Ngf
Gene Alias Ngfb
Gene Symbols Ngf
Host Species Sheep
Immunogen Native purified mouse Ngf.
Quantity 250 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18049
Test Specificity A cross reactivity of less than 1% to recombinant human BDNF, NT3, NT4/5 by ELISA has been shown.
Target Species Avian, Human, Mouse, Rat
Content And Storage Store at 4°C on dry atmosphere. After reconstitution with deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.