Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NTF3, Sheep, Polyclonal Antibody, Abnova™

Catalog No. 89117013 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-117-013 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-117-013 Supplier Abnova Corporation Supplier No. PAB14343
Add to Cart
Add to Cart

Sheep polyclonal antibody raised against synthetic peptide of NTF3.

Sequence: YAEHKSHRGEY

Specifications

Antigen NTF3
Applications Dot Blot, ELISA, Immunohistochemistry, Inhibition Assays, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Sheep polyclonal antibody raised against synthetic peptide of NTF3.
Dilution Western Blot (1-10 ug/mL) The optimal working dilution should be determined by the end user.
Formulation Lyophilized from PBS
Gene Alias HDNF/MGC129711/NGF-2/NGF2/NT3
Gene Symbols NTF3
Host Species Sheep
Immunogen A synthetic peptide (conjugated with BSA) corresponding to human, mouse and rat NTF3/Ntf3.
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4908/18205/81737
Test Specificity A cross reactivity of less than 0.1% to mouse NGF, recombinant human BDNF and NT4/5 has been shown by dot blot.
Target Species Chicken, Human, Mouse, Rat
Content And Storage Store at 4°C on dry atmosphere. After reconstitution with deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.